Assay Details
| Gene Symbol: Ksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013571.2 | NP_038599.1 | 7 - 8 | 1164 | 80 | |
| GenBank mRNA | AK036566.1 | - | 7 - 8 | 1220 | 80 | |
| AK137532.1 | - | 6 - 7 | 901 | 80 | ||
| AK164833.1 | - | 7 - 8 | 1286 | 80 | ||
| U43585.1 | - | 7 - 8 | 1171 | 80 | ||
| X81634.1 | - | 7 - 8 | 946 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ksr1
Genomic Map