Assay Details
| Gene Symbol: Ksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013571.2 | NP_038599.1 | 14 - 15 | 2080 | 93 | |
| GenBank mRNA | AK137532.1 | - | 13 - 14 | 1817 | 93 | |
| U43585.1 | - | 14 - 15 | 2087 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ksr1
Genomic Map