Assay Details
| Gene Symbol: Irf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013674.1 | NP_038702.1 | 7 - 8 | 1168 | 62 | |
| GenBank mRNA | AK089319.1 | - | 7 - 8 | 1187 | 62 | |
| AK089819.1 | - | 7 - 8 | 1190 | 62 | ||
| AK154138.1 | - | 7 - 8 | 1192 | 62 | ||
| AK154315.1 | - | 7 - 8 | 1180 | 62 | ||
| BC137713.1 | - | 7 - 8 | 1156 | 8860732 | 62 | |
| BC137714.1 | - | 7 - 8 | 1156 | 8860733 | 62 | |
| U11692.1 | - | 7 - 8 | 1168 | 62 | ||
| U34307.1 | - | 7 - 8 | 1162 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Irf4
Genomic Map