Assay Details
| Gene Symbol: Itgb1bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013712.2 | NP_038740.1 | 4 - 5 | 377 | 68 | |
| GenBank mRNA | AF140691.1 | - | 4 - 5 | 321 | 68 | |
| AK003906.1 | - | 4 - 5 | 378 | 68 | ||
| BC030035.1 | - | 4 - 5 | 352 | 1346742 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P298088 | AK003906.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P298088 | BC030035.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P298088 | NM_013712.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P298088 | AF140691.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P298088 | AK003906.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P298088 | BC030035.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P298088 | NM_013712.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P298088 | AF140691.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419726 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 959958 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 356353 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 595286 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178929 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P298088 | AK003906.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P298088 | BC030035.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P298088 | NM_013712.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P298088 | AF140691.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P298088 | AK003906.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P298088 | BC030035.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P298088 | NM_013712.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P298088 | AF140691.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P298088 | AK003906.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P298088 | BC030035.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P298088 | NM_013712.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P298088 | AF140691.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P298088 | AK003906.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P298088 | BC030035.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P298088 | NM_013712.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P298088 | AF140691.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32564170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32564180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32564176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32564181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32564169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32564178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32564173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32564175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32564166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32564182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32564172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32564177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32564165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32564168 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178929 | - | - |
| Whole Mouse Genome G4122A | A_52_P298088 | AK003906.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P298088 | BC030035.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P298088 | NM_013712.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P298088 | AF140691.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P298088 | AK003906.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P298088 | BC030035.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P298088 | NM_013712.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P298088 | AF140691.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124483 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178929 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14983323 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14983337 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14983314 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14983336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32564185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14983319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14554563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14983342 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564168 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14554564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32564205 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10601186 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601189 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601181 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601183 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601188 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601184 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601185 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601190 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601180 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601187 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601179 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601182 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601191 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423238_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10601178 | - | - |
| Mouse Genome 430 2.0 Array | 1423238_at | - | - |
| Mouse Genome 430 2.0 Array | 1423238_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26549_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26549_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00006488.mm.1 | NM_013712.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006488.mm.1 | NM_013712.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006488.mm.1 | TC0X00000975.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423238_PM_at | - | - |
| Mouse Expression Set 430 | 1423238_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423238_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423238_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423238_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423238_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 318088 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6200129 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6200129 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006200129 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1253770 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253770 | - | - |
| mouse-6 v1.1 (Illumina) | scl54739.8.5_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6200129 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl54739.8.5_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6200129 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2853166 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2853166 | - | - |

Itgb1bp2
Genomic Map