Assay Details
| Gene Symbol: Dbf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190717.1 | NP_001177646.1 | 9 - 10 | 1008 | 63 | |
| NM_013726.3 | NP_038754.1 | 9 - 10 | 1011 | 63 | ||
| GenBank mRNA | AJ003132.1 | - | 9 - 10 | 901 | 63 | |
| AK014480.1 | - | 9 - 10 | 977 | 63 | ||
| AK083650.1 | - | 6 - 7 | 386 | 63 | ||
| AK132403.1 | - | 9 - 10 | 973 | 63 | ||
| AK163408.1 | - | 7 - 8 | 587 | 63 | ||
| BC069983.1 | - | 9 - 10 | 959 | 63 | ||
| BC138709.1 | - | 9 - 10 | 829 | 8861731 | 63 | |
| BC138710.1 | - | 9 - 10 | 829 | 8861732 | 63 | |
| BC145392.1 | - | 9 - 10 | 826 | 9053950 | 63 | |
More Information
Set Membership: |
|

Dbf4
Genomic Map