Assay Details
| Gene Symbol: Barx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013800.2 | NP_038828.2 | 2 - 3 | 694 | 87 | |
| GenBank mRNA | BC012684.1 | - | 2 - 3 | 685 | 3980774 | 87 |
| L77900.1 | - | 2 - 3 | 492 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Barx2
Genomic Map