Assay Details
| Gene Symbol: Fbxo18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015792.1 | NP_056607.1 | 15 - 16 | 2413 | 81 | |
| GenBank mRNA | AF184275.1 | - | 13 - 14 | 1907 | 81 | |
| AK083118.1 | - | 15 - 16 | 2758 | 81 | ||
| AK084645.1 | - | 15 - 16 | 2350 | 81 | ||
| AK147829.1 | - | 15 - 16 | 2397 | 81 | ||
| AK154357.1 | - | 15 - 16 | 2436 | 81 | ||
| AK158937.1 | - | 15 - 16 | 2464 | 81 | ||
| AK181430.1 | - | 1 - 2 | 44 | 81 | ||
| AK208346.1 | - | 1 - 2 | 44 | 81 | ||
| AK211398.1 | - | 2 - 3 | 145 | 81 | ||
| BC004036.1 | - | 12 - 13 | 1499 | 81 | ||
| BC031393.1 | - | 15 - 16 | 2413 | 4948804 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo18
Genomic Map