Assay Details
| Gene Symbol: Cntfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136056.2 | NP_001129528.1 | 6 - 7 | 831 | 60 | |
| NM_016673.2 | NP_057882.2 | 6 - 7 | 810 | 60 | ||
| GenBank mRNA | AB154246.1 | - | 4 - 5 | 606 | 60 | |
| AF068615.1 | - | 6 - 7 | 761 | 60 | ||
| AK137026.1 | - | 6 - 7 | 720 | 60 | ||
| BC046974.1 | - | 6 - 7 | 800 | 6389656 | 60 | |
| BC050928.1 | - | 6 - 7 | 828 | 6415187 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cntfr
Genomic Map