Assay Details
Gene Symbol: Cul3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016716.4 | NP_057925.1 | 11 - 12 | 1729 | 70 | |
GenBank mRNA | AF129738.1 | - | 11 - 12 | 1619 | 70 | |
AK129174.1 | - | 12 - 13 | 2117 | 70 | ||
BC027304.1 | - | 11 - 12 | 1754 | 3981170 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
