Assay Details
| Gene Symbol: Ddah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190449.1 | NP_001177378.1 | 4 - 5 | 769 | 65 | |
| NM_016765.3 | NP_058045.1 | 4 - 5 | 769 | 65 | ||
| GenBank mRNA | AF004106.1 | - | 4 - 5 | 703 | 65 | |
| BC003328.1 | - | 4 - 5 | 719 | 3158175 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddah2
Genomic Map