Assay Details
| Gene Symbol: Deaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282072.1 | NP_001269001.1 | 3 - 4 | 676 | 115 | |
| NM_001282073.1 | NP_001269002.1 | 4 - 5 | 600 | 115 | ||
| NM_001282076.1 | NP_001269005.1 | 4 - 5 | 633 | 115 | ||
| NM_016874.3 | NP_058570.1 | 3 - 4 | 676 | 115 | ||
| GenBank mRNA | AF102818.1 | - | 3 - 4 | 536 | 115 | |
| AI430175.1 | - | 2 - 3 | 158 | 115 | ||
| AK082115.1 | - | 3 - 4 | 529 | 115 | ||
| AK144333.1 | - | 3 - 4 | 445 | 115 | ||
| AK146546.1 | - | 3 - 4 | 553 | 115 | ||
| AK155836.1 | - | 3 - 4 | 642 | 115 | ||
| AK158586.1 | - | 4 - 5 | 485 | 115 | ||
| BC046399.1 | - | 4 - 5 | 578 | 5063995 | 115 | |
| FJ377318.1 | - | 4 - 5 | 493 | 115 | ||
| FJ377319.1 | - | 3 - 4 | 521 | 115 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001282072.1,NM_001282076.1 transcripts.
Set Membership: |
|

Deaf1
Genomic Map