| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700038086.mm.1 |
NM_001282072.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038086.mm.1 |
NM_001282076.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038086.mm.1 |
TC0700004533.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700038088.mm.1 |
NM_001282072.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038088.mm.1 |
NM_001282073.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038088.mm.1 |
NM_001282076.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038088.mm.1 |
NM_016874.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038088.mm.1 |
TC0700004533.mm.1 |
MTA TC |