Assay Details
| Gene Symbol: Deaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282072.1 | NP_001269001.1 | 9 - 10 | 1456 | 110 | |
| NM_001282073.1 | NP_001269002.1 | 9 - 10 | 1210 | 110 | ||
| NM_001282076.1 | NP_001269005.1 | 10 - 11 | 1413 | 110 | ||
| NM_016874.3 | NP_058570.1 | 8 - 9 | 1286 | 110 | ||
| GenBank mRNA | AF102818.1 | - | 8 - 9 | 1146 | 110 | |
| AK082115.1 | - | 8 - 9 | 1139 | 110 | ||
| AK144333.1 | - | 9 - 10 | 1225 | 110 | ||
| AK146546.1 | - | 8 - 9 | 1163 | 110 | ||
| AK155836.1 | - | 9 - 10 | 1422 | 110 | ||
| AK158586.1 | - | 9 - 10 | 1095 | 110 | ||
| BC046399.1 | - | 9 - 10 | 1188 | 5063995 | 110 | |
| CD353366.1 | - | 2 - 3 | 222 | 110 | ||
| FJ377318.1 | - | 10 - 11 | 1273 | 110 | ||
| FJ377319.1 | - | 8 - 9 | 1131 | 110 | ||
| W53084.1 | - | 2 - 3 | 199 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Deaf1
Genomic Map