Assay Details
| Gene Symbol: Deaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282072.1 | NP_001269001.1 | 11 - 12 | 1832 | 56 | |
| NM_016874.3 | NP_058570.1 | 10 - 11 | 1662 | 56 | ||
| GenBank mRNA | AF102818.1 | - | 10 - 11 | 1522 | 56 | |
| AK144333.1 | - | 11 - 12 | 1601 | 56 | ||
| AK146546.1 | - | 10 - 11 | 1539 | 56 | ||
| CD353366.1 | - | 4 - 5 | 598 | 56 | ||
| FJ377319.1 | - | 10 - 11 | 1507 | 56 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | CO042018.1 | - | Chr. - - [-] on Build | 56 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Deaf1
Genomic Map