Assay Details
| Gene Symbol: Fanca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016925.3 | NP_058621.2 | 12 - 13 | 1097 | 75 | |
| GenBank mRNA | AF208116.1 | - | 12 - 13 | 1099 | 75 | |
| AF208117.1 | - | 9 - 10 | 780 | 75 | ||
| AF208118.1 | - | 12 - 13 | 1099 | 75 | ||
| AK084990.1 | - | 11 - 12 | 1205 | 75 | ||
| AK166112.1 | - | 12 - 13 | 1082 | 75 | ||
| BC150705.1 | - | 12 - 13 | 1095 | 9087616 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fanca
Genomic Map