Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100034507.mm.1 |
NM_016958.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034507.mm.1 |
TC1100003760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034508.mm.1 |
NM_016958.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034508.mm.1 |
TC1100003760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034506.mm.1 |
NM_016958.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034506.mm.1 |
TC1100003760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034509.mm.1 |
NM_016958.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034509.mm.1 |
TC1100003760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034511.mm.1 |
NM_016958.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034511.mm.1 |
TC1100003760.mm.1 |
MTA TC |