Assay Details
| Gene Symbol: Gucy1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161796.1 | NP_001155268.1 | 1 - 2 | 142 | 73 | |
| NM_017469.4 | NP_059497.1 | 1 - 2 | 142 | 73 | ||
| GenBank mRNA | AF020339.1 | - | 1 - 2 | 116 | 73 | |
| AF297083.1 | - | 1 - 2 | 116 | 73 | ||
| AK139406.1 | - | 1 - 2 | 105 | 73 | ||
| AK147128.1 | - | 1 - 2 | 77 | 73 | ||
| BC050945.1 | - | 1 - 2 | 120 | 6822142 | 73 | |
| BC056995.1 | - | 1 - 2 | 120 | 6822142 | 73 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF020339.1,AF297083.1,AK139406.1,BC050945.1,BC056995.1,NM_001161796.1,NM_017469.4 transcripts.
Set Membership: |
|

Gucy1b3
Genomic Map