Assay Details
| Gene Symbol: Gucy1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161796.1 | NP_001155268.1 | 6 - 7 | 867 | 74 | |
| NM_017469.4 | NP_059497.1 | 6 - 7 | 867 | 74 | ||
| GenBank mRNA | AF020339.1 | - | 6 - 7 | 841 | 74 | |
| AF297083.1 | - | 6 - 7 | 841 | 74 | ||
| AK139406.1 | - | 6 - 7 | 830 | 74 | ||
| AK147128.1 | - | 6 - 7 | 802 | 74 | ||
| BC050945.1 | - | 6 - 7 | 845 | 6822142 | 74 | |
| BC056995.1 | - | 6 - 7 | 845 | 6822142 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gucy1b3
Genomic Map