Assay Details
| Gene Symbol: Crlf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018827.2 | NP_061297.1 | 5 - 6 | 981 | 84 | |
| GenBank mRNA | AB040038.1 | - | 5 - 6 | 981 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crlf1
Genomic Map