Assay Details
| Gene Symbol: Camkk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018883.2 | NP_061371.2 | 2 - 3 | 523 | 61 | |
| GenBank mRNA | AF117384.1 | - | 2 - 3 | 478 | 61 | |
| AK144483.1 | - | 2 - 3 | 524 | 61 | ||
| BC017529.1 | - | 2 - 3 | 488 | 4924656 | 61 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Camkk1
Genomic Map