Assay Details
| Gene Symbol: Cyp39a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285947.1 | NP_001272876.1 | 7 - 8 | 1154 | 79 | |
| NM_001285948.1 | NP_001272877.1 | 8 - 9 | 1304 | 79 | ||
| NM_018887.4 | NP_061375.1 | 8 - 9 | 1304 | 79 | ||
| GenBank mRNA | AF237981.1 | - | 8 - 9 | 1153 | 79 | |
| AK031733.1 | - | 8 - 9 | 1252 | 79 | ||
| BC021418.1 | - | 8 - 9 | 1175 | 5025117 | 79 | |
| BC037663.1 | - | 7 - 8 | 1003 | 5352740 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp39a1
Genomic Map