Assay Details
| Gene Symbol: Avpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019404.2 | NP_062277.1 | 1 - 2 | 169 | 80 | |
| GenBank mRNA | AJ310753.1 | - | 1 - 2 | 169 | 80 | |
| BC104729.1 | - | 1 - 2 | 161 | 40046723 | 80 | |
| BC104731.1 | - | 1 - 2 | 161 | 40046726 | 80 | |
| BC105662.1 | - | 1 - 2 | 161 | 40046727 | 80 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AJ310753.1,BC104729.1,BC104731.1,BC105662.1,NM_019404.2 transcripts.
Set Membership: |
|

Avpr2
Genomic Map