Assay Details
| Gene Symbol: Elovl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039175.2 | NP_001034264.1 | 1 - 2 | 122 | 82 | |
| NM_019422.3 | NP_062295.1 | 1 - 2 | 122 | 82 | ||
| GenBank mRNA | AF170907.1 | - | 1 - 2 | 115 | 82 | |
| AK003743.1 | - | 1 - 2 | 91 | 82 | ||
| AK135348.1 | - | 1 - 2 | 97 | 82 | ||
| AK171217.1 | - | 1 - 2 | 108 | 82 | ||
| BC096673.1 | - | 1 - 2 | 101 | 5143739 | 82 | |
| BY141978.1 | - | 1 - 2 | 122 | 82 | ||
| CX199666.1 | - | 1 - 2 | 95 | 82 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF170907.1,AK003743.1,AK135348.1,AK171217.1,BC096673.1,NM_001039175.2,NM_019422.3 transcripts.
Set Membership: |
|

Elovl1
Genomic Map