Assay Details
| Gene Symbol: Elovl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039175.2 | NP_001034264.1 | 5 - 6 | 425 | 78 | |
| NM_001039176.2 | NP_001034265.1 | 6 - 7 | 623 | 78 | ||
| NM_019422.3 | NP_062295.1 | 6 - 7 | 616 | 78 | ||
| GenBank mRNA | AF104033.1 | - | 3 - 4 | 182 | 78 | |
| AF170907.1 | - | 6 - 7 | 609 | 78 | ||
| AK003743.1 | - | 5 - 6 | 394 | 78 | ||
| AK135348.1 | - | 6 - 7 | 591 | 78 | ||
| AK156003.1 | - | 6 - 7 | 2217 | 78 | ||
| AK171217.1 | - | 6 - 7 | 602 | 78 | ||
| BC006735.1 | - | 6 - 7 | 623 | 3156004 | 78 | |
| BC096673.1 | - | 5 - 6 | 404 | 5143739 | 78 | |
| CN455861.1 | - | 4 - 5 | 360 | 78 | ||
| CX199666.1 | - | 6 - 7 | 589 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Elovl1
Genomic Map