Assay Details
| Gene Symbol: Bace2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019517.4 | NP_062390.3 | 4 - 5 | 809 | 144 | |
| GenBank mRNA | AF216310.1 | - | 4 - 5 | 760 | 144 | |
| AK052309.1 | - | 4 - 5 | 796 | 144 | ||
| AK078770.1 | - | 4 - 5 | 776 | 144 | ||
| AK165036.1 | - | 4 - 5 | 745 | 144 | ||
| AK184226.1 | - | 1 - 2 | 49 | 144 | ||
| AK185329.1 | - | 1 - 2 | 49 | 144 | ||
| BC120773.1 | - | 4 - 5 | 775 | 40129696 | 144 | |
| BC131947.1 | - | 4 - 5 | 775 | 40130224 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bace2
Genomic Map