Assay Details
| Gene Symbol: Grasp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019518.3 | NP_062391.3 | 3 - 4 | 441 | 118 | |
| GenBank mRNA | AB041603.1 | - | 3 - 4 | 439 | 118 | |
| AF236099.1 | - | 3 - 4 | 404 | 118 | ||
| BC046307.1 | - | 3 - 4 | 409 | 6306542 | 118 | |
| BY730445.1 | - | 3 - 4 | 460 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grasp
Genomic Map