Assay Details
| Gene Symbol: Extl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF224461.1 | - | 4 - 5 | 1137 | 79 | |
| AK172619.1 | - | 4 - 5 | 1907 | 79 | ||
| BC120890.1 | - | 4 - 5 | 1137 | 40061413 | 79 | |
| BC120891.1 | - | 4 - 5 | 1137 | 40061414 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Extl1
Genomic Map