Assay Details
| Gene Symbol: Extl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019578.2 | NP_062524.2 | 5 - 6 | 2039 | 71 | |
| GenBank mRNA | AF224461.1 | - | 5 - 6 | 1271 | 71 | |
| AK085075.1 | - | 5 - 6 | 2041 | 71 | ||
| AK161915.1 | - | 5 - 6 | 1112 | 71 | ||
| AK164184.1 | - | 4 - 5 | 1929 | 71 | ||
| AK172619.1 | - | 5 - 6 | 2041 | 71 | ||
| AK218323.1 | - | 1 - 2 | 69 | 71 | ||
| BC120890.1 | - | 5 - 6 | 1271 | 40061413 | 71 | |
| BC120891.1 | - | 5 - 6 | 1271 | 40061414 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Extl1
Genomic Map