Assay Details
| Gene Symbol: Rapgef4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204165.1 | NP_001191094.1 | 8 - 9 | 770 | 76 | |
| NM_001204166.1 | NP_001191095.1 | 5 - 6 | 662 | 76 | ||
| NM_019688.2 | NP_062662.1 | 7 - 8 | 716 | 76 | ||
| GenBank mRNA | AB021132.1 | - | 8 - 9 | 745 | 76 | |
| AB445011.1 | - | 5 - 6 | 654 | 76 | ||
| AF115480.1 | - | 7 - 8 | 644 | 76 | ||
| AK220281.1 | - | 8 - 9 | 770 | 76 | ||
| BC139822.1 | - | 8 - 9 | 702 | 40141811 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rapgef4
Genomic Map