Assay Details
| Gene Symbol: Fads2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019699.1 | NP_062673.1 | 4 - 5 | 692 | 76 | |
| GenBank mRNA | AF126798.1 | - | 4 - 5 | 692 | 76 | |
| AK083282.1 | - | 659 | 76 | |||
| AK143524.1 | - | 710 | 76 | |||
| BC057189.1 | - | 4 - 5 | 680 | 5137343 | 76 | |
| HQ264057.1 | - | 4 - 5 | 585 | 76 | ||
| HQ264058.1 | - | 4 - 5 | 585 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fads2
Genomic Map