Assay Details
| Gene Symbol: Gps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019726.3 | NP_062700.2 | 9 - 10 | 1052 | 140 | |
| GenBank mRNA | AF153696.1 | - | 10 - 11 | 992 | 140 | |
| BC011317.1 | - | 9 - 10 | 1052 | 3979292 | 140 | |
| BC138879.1 | - | 9 - 10 | 1008 | 8861901 | 140 | |
| BC138880.1 | - | 9 - 10 | 1009 | 8861902 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27897910 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27897906 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27897911 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27897905 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27897907 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27897908 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27897909 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957583 | NM_019726.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957584 | BC138879.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957584 | BC138880.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957584 | NM_019726.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957584 | AF153696.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957584 | BC011317.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852611 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 582364 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957584 | BC138879.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957584 | BC138880.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957584 | NM_019726.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957584 | AF153696.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957584 | BC011317.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957583 | NM_019726.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27897912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27897925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27897906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27897908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27897918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27897921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27897909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27897913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27897923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27897916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27897915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27897910 | - | - |
| Whole Mouse Genome G4122A | A_51_P320786 | AF153696.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320782 | BC138879.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320782 | BC138880.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320782 | NM_019726.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320782 | AF153696.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320782 | BC011317.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116708 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138311 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09457832 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27897910 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27897906 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27897908 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27897909 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27897907 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27897905 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27897911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27897923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27897925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27897923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27897912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27897921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27897915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27897916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09055089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27897913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09457832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27897918 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10377654 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377659 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377661 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377655 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377653 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377658 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377656 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377660 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377657 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417796_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10377652 | - | - |
| Mouse Genome 430 2.0 Array | 1417796_at | - | - |
| Mouse Genome 430 2.0 Array | 1417796_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56310_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56310_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100007981.mm.1 | NM_019726.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100007981.mm.1 | NM_019726.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100007981.mm.1 | TC1100000929.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100007980.mm.1 | NM_019726.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100007980.mm.1 | NM_019726.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100007980.mm.1 | TC1100000929.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417796_at | - | - |
| Mouse Expression Set 430 | 1417796_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417796_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417796_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417796_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417796_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 719621 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5910672 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5910672 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005910672 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2730784 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2730784 | - | - |
| mouse-6 v1.1 (Illumina) | scl056310.10_41-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5910672 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl056310.10_41-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5910672 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2730784 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2730784 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl056310.10_41-S | - | - |

Gps2
Genomic Map