Assay Details
| Gene Symbol: Dnajc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019795.4 | NP_062769.2 | 7 - 8 | 1315 | 68 | |
| GenBank mRNA | AB028861.1 | - | 7 - 8 | 763 | 68 | |
| AK011384.1 | - | 2 - 3 | 173 | 68 | ||
| AK076032.1 | - | 7 - 8 | 786 | 68 | ||
| AK145506.1 | - | 7 - 8 | 777 | 68 | ||
| AK145739.1 | - | 7 - 8 | 777 | 68 | ||
| AK166872.1 | - | 7 - 8 | 777 | 68 | ||
| AY323828.1 | - | 7 - 8 | 751 | 68 | ||
| BC023681.1 | - | 7 - 8 | 959 | 5343384 | 68 | |
| BC055729.1 | - | 7 - 8 | 1314 | 6414499 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc7
Genomic Map