Assay Details
| Gene Symbol: Dnajc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019795.4 | NP_062769.2 | 10 - 11 | 1649 | 85 | |
| GenBank mRNA | AB028861.1 | - | 10 - 11 | 1097 | 85 | |
| AK011384.1 | - | 5 - 6 | 507 | 85 | ||
| AK011875.1 | - | 10 - 11 | 1088 | 85 | ||
| AK076032.1 | - | 10 - 11 | 1120 | 85 | ||
| AK145506.1 | - | 10 - 11 | 1111 | 85 | ||
| AK145739.1 | - | 10 - 11 | 1111 | 85 | ||
| AK166872.1 | - | 10 - 11 | 1111 | 85 | ||
| AY323828.1 | - | 10 - 11 | 1085 | 85 | ||
| BC023681.1 | - | 10 - 11 | 1293 | 5343384 | 85 | |
| BC055729.1 | - | 10 - 11 | 1648 | 6414499 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc7
Genomic Map