Assay Details
| Gene Symbol: Anapc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019805.4 | NP_062779.3 | 3 - 4 | 538 | 63 | |
| GenBank mRNA | AF076607.2 | - | 3 - 4 | 502 | 63 | |
| AK030775.1 | - | 3 - 4 | 503 | 63 | ||
| AK049968.1 | - | 3 - 4 | 477 | 63 | ||
| AK137286.1 | - | 3 - 4 | 502 | 63 | ||
| AK150764.1 | - | 3 - 4 | 502 | 63 | ||
| AK150891.1 | - | 3 - 4 | 502 | 63 | ||
| AK153114.1 | - | 3 - 4 | 502 | 63 | ||
| AK159523.1 | - | 3 - 4 | 470 | 63 | ||
| BC006635.1 | - | 3 - 4 | 491 | 3482261 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anapc7
Genomic Map