Assay Details
| Gene Symbol: Anapc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019805.4 | NP_062779.3 | 9 - 10 | 1488 | 68 | |
| GenBank mRNA | AF076607.2 | - | 9 - 10 | 1452 | 68 | |
| AK030775.1 | - | 9 - 10 | 1453 | 68 | ||
| AK049968.1 | - | 9 - 10 | 1427 | 68 | ||
| AK137286.1 | - | 9 - 10 | 1452 | 68 | ||
| AK150764.1 | - | 9 - 10 | 1452 | 68 | ||
| AK150891.1 | - | 9 - 10 | 1452 | 68 | ||
| AK153114.1 | - | 9 - 10 | 1452 | 68 | ||
| AK159523.1 | - | 9 - 10 | 1420 | 68 | ||
| AK164202.1 | - | 9 - 10 | 1457 | 68 | ||
| BC006635.1 | - | 9 - 10 | 1441 | 3482261 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anapc7
Genomic Map