Assay Details
| Gene Symbol: Dnajb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019965.2 | NP_064349.2 | 6 - 7 | 922 | 67 | |
| GenBank mRNA | AB028860.1 | - | 6 - 7 | 895 | 67 | |
| AK038149.1 | - | 5 - 6 | 803 | 67 | ||
| AK082546.1 | - | 6 - 7 | 872 | 67 | ||
| AK138495.1 | - | 6 - 7 | 852 | 67 | ||
| AK166224.1 | - | 6 - 7 | 909 | 67 | ||
| AK172274.1 | - | 6 - 7 | 909 | 67 | ||
| BC034162.1 | - | 6 - 7 | 922 | 5043868 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnajb12
Genomic Map