Assay Details
| Gene Symbol: Stap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019992.3 | NP_064376.1 | 3 - 4 | 314 | 147 | |
| GenBank mRNA | AB036058.1 | - | 3 - 4 | 304 | 147 | |
| AK041474.1 | - | 3 - 4 | 315 | 147 | ||
| AK153661.1 | - | 3 - 4 | 393 | 147 | ||
| AK162655.1 | - | 3 - 4 | 396 | 147 | ||
| BC145828.1 | - | 3 - 4 | 329 | 40131131 | 147 | |
| BC145830.1 | - | 3 - 4 | 330 | 40131133 | 147 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stap1
Genomic Map