Assay Details
| Gene Symbol: Stap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019992.3 | NP_064376.1 | 6 - 7 | 673 | 88 | |
| GenBank mRNA | AB036058.1 | - | 6 - 7 | 88 | ||
| AK041474.1 | - | 6 - 7 | 674 | 88 | ||
| AK153661.1 | - | 6 - 7 | 752 | 88 | ||
| AK162655.1 | - | 6 - 7 | 755 | 88 | ||
| BC145828.1 | - | 6 - 7 | 688 | 40131131 | 88 | |
| BC145830.1 | - | 6 - 7 | 689 | 40131133 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stap1
Genomic Map