Assay Details
| Gene Symbol: C1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020558.3 | NP_065583.2 | 4 - 5 | 308 | 98 | |
| GenBank mRNA | AK028702.1 | - | 4 - 5 | 315 | 98 | |
| AK035169.1 | - | 4 - 5 | 309 | 98 | ||
| BC005436.1 | - | 4 - 5 | 304 | 3591682 | 98 | |
| X95591.1 | - | 4 - 5 | 335 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
maturation of 5.8S rRNA
transcription, DNA-templated
regulation of transcription, DNA-templated
rRNA processing
apoptotic process
negative regulation of transcription, DNA-templated
DNA binding
transcription corepressor activity
RNA binding
protein binding
ligand-dependent nuclear receptor binding
maturation of 5.8S rRNA
transcription, DNA-templated
regulation of transcription, DNA-templated
rRNA processing
apoptotic process
negative regulation of transcription, DNA-templated
DNA binding
transcription corepressor activity
RNA binding
protein binding
ligand-dependent nuclear receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27615063 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27615064 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010946 | AK035169.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010946 | NM_020558.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 658121 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285764 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826083 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 832022 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149708 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 80027 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156669 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161882 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010946 | AK035169.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010946 | NM_020558.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P150495 | AK035169.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P150495 | NM_020558.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P150495 | AK028702.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27615055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27615057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27615068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27615071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27615059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27615079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27615054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27615074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27615063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27615061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27615073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27615067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27615064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27615077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27615078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27615065 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161882 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156669 | - | - |
| Whole Mouse Genome G4122A | A_52_P339543 | AK035169.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P339543 | BC005436.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P339543 | X95591.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P339543 | NM_020558.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P339543 | AK028702.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P150495 | AK035169.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P150495 | NM_020558.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P150495 | AK028702.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130206 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156669 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161882 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09398968 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09398967 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09156562 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27615063 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27615064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27615166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09503547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09135855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615077 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09464628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09154115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09403562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09440084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09154112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09135860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09464621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27615183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09403561 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10374446 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374445 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374447 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374443 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374444 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416887_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416886_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448505_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10374442 | - | - |
| Mouse Genome 430 2.0 Array | 1416886_at | - | - |
| Mouse Genome 430 2.0 Array | 1416886_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416887_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448505_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448505_at | - | - |
| Mouse Genome 430 2.0 Array | 1416887_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57316_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57316_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100001552.mm.1 | NM_020558.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100001552.mm.1 | TC1100000170.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100001550.mm.1 | NM_020558.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100001550.mm.1 | TC1100000170.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100001551.mm.1 | NM_020558.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100001551.mm.1 | TC1100000170.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100001549.mm.1 | NM_020558.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100001549.mm.1 | TC1100000170.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448505_at | - | - |
| Mouse Expression Set 430 | 1448505_PM_at | - | - |
| Mouse Expression Set 430 | 1416887_at | - | - |
| Mouse Expression Set 430 | 1416886_PM_at | - | - |
| Mouse Expression Set 430 | 1416886_at | - | - |
| Mouse Expression Set 430 | 1416887_PM_at | - | - |
| Murine Genome U74v2 Set | 95479_at | - | - |
| Murine Genome U74v2 Set | 95479_PM_at | - | - |
| Murine Genome U74v2 Set | 130426_r_at | - | - |
| Murine Genome U74v2 Set | 130426_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416886_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416887_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448505_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448505_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416886_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416887_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416887_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448505_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448505_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416886_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416886_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416887_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 819122 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160140 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3160140 | - | - |
| mouse-6 v1.1 (Illumina) | GI_31542250-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3160140 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160140 | - | - |

C1d
Genomic Map