Assay Details
| Gene Symbol: Jmy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021310.3 | NP_067285.2 | 2 - 3 | 1677 | 64 | |
| GenBank mRNA | AB221690.1 | - | 2 - 3 | 1175 | 64 | |
| AF201390.1 | - | 2 - 3 | 1301 | 64 | ||
| AK141957.1 | - | 2 - 3 | 1678 | 64 | ||
| BC020052.1 | - | 2 - 3 | 1602 | 64 | ||
| BC069906.1 | - | 2 - 3 | 1531 | 64 | ||
| BC090835.1 | - | 2 - 3 | 1558 | 5705427 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jmy
Genomic Map