Assay Details
| Gene Symbol: Gucy1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021896.5 | NP_068696.2 | 1 - 2 | 207 | 68 | |
| GenBank mRNA | AF297082.1 | - | 1 - 2 | 182 | 68 | |
| AK004815.1 | - | 1 - 2 | 141 | 68 | ||
| AK028558.1 | - | 1 - 2 | 130 | 68 | ||
| AK031305.1 | - | 1 - 2 | 120 | 68 | ||
| BC057327.1 | - | 1 - 2 | 117 | 6825960 | 68 | |
| BY150724.1 | - | 1 - 2 | 208 | 68 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF297082.1,AK004815.1,AK028558.1,BC057327.1,NM_021896.5 transcripts.
Set Membership: |
|

Gucy1a3
Genomic Map