Assay Details
| Gene Symbol: Eif4enif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166547.1 | NP_001160019.1 | 17 - 18 | 2778 | 70 | |
| NM_001166548.1 | NP_001160020.1 | 16 - 17 | 2710 | 70 | ||
| NM_001166549.1 | NP_001160021.1 | 16 - 17 | 2814 | 70 | ||
| NM_023743.2 | NP_076232.2 | 17 - 18 | 2793 | 70 | ||
| GenBank mRNA | AB031388.1 | - | 17 - 18 | 2614 | 70 | |
| AK012082.1 | - | 7 - 8 | 1197 | 70 | ||
| AK041598.1 | - | 16 - 17 | 2672 | 70 | ||
| AK088425.1 | - | 12 - 13 | 1880 | 70 | ||
| AK165678.1 | - | 17 - 18 | 2780 | 70 | ||
| BC033410.1 | - | 17 - 18 | 2628 | 5359290 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif4enif1
Genomic Map