Assay Details
| Gene Symbol: Stambp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024239.2 | NP_077201.1 | 8 - 9 | 1227 | 81 | |
| GenBank mRNA | AB010123.1 | - | 8 - 9 | 1174 | 81 | |
| AK017600.1 | - | 8 - 9 | 1182 | 81 | ||
| AK019907.1 | - | 8 - 9 | 1177 | 81 | ||
| AK046995.1 | - | 8 - 9 | 1178 | 81 | ||
| AK136961.1 | - | 8 - 9 | 1778 | 81 | ||
| AK139391.1 | - | 8 - 9 | 1202 | 81 | ||
| AK168810.1 | - | 8 - 9 | 1200 | 81 | ||
| AK220541.1 | - | 6 - 7 | 7458 | 81 | ||
| BC003497.1 | - | 8 - 9 | 1170 | 3153829 | 81 | |
| BC006939.1 | - | 8 - 9 | 1225 | 2649718 | 81 | |
| BC025111.1 | - | 8 - 9 | 1169 | 4194332 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stambp
Genomic Map