Assay Details
| Gene Symbol: Sh3bp5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK086088.1 | - | 4 - 5 | 850 | 66 | |
| AK144928.1 | - | 4 - 5 | 849 | 66 | ||
| AK150129.1 | - | 4 - 5 | 837 | 66 | ||
| AK156171.1 | - | 4 - 5 | 890 | 66 | ||
| AK159202.1 | - | 4 - 5 | 880 | 66 | ||
| AK166491.1 | - | 4 - 5 | 850 | 66 | ||
| AK167203.1 | - | 4 - 5 | 964 | 66 | ||
| AK172333.1 | - | 4 - 5 | 890 | 66 | ||
| BC003251.1 | - | 3 - 4 | 511 | 3500047 | 66 | |
| BC094418.1 | - | 4 - 5 | 872 | 30702568 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27830412 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149274 | BC094418.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149274 | AK122543.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149274 | BC003251.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149274 | BC013661.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149274 | AK167203.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095688 | BC003251.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095688 | AK086088.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095688 | AK144928.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095688 | NM_001161338.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095688 | NM_024480.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662762 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 629745 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680769 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 194346 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 667189 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616680 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718011 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 73891 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76167 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110701 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114639 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149274 | BC094418.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149274 | AK122543.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149274 | BC003251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149274 | BC013661.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149274 | AK167203.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095688 | BC003251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095688 | AK086088.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095688 | AK144928.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095688 | NM_001161338.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095688 | NM_024480.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27830416 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114639 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02335526 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07357155 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110701 | - | - |
| Whole Mouse Genome G4122A | A_51_P115988 | AK122543.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P115988 | BC003251.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P115988 | BC013661.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P115988 | AK172333.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P115988 | AK156171.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137248 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110701 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114639 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09019195 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09019171 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09019194 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09019179 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09019166 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27830412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27830460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09019195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09386752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09093140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08992831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09093144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09019186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09251437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09019169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09399289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09149102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09084988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09053354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08992832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09093149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27830432 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10376361 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376363 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376360 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376364 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376365 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376362 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376359 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416223_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10376358 | - | - |
| Mouse Genome 430 2.0 Array | 1416223_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416223_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 79566_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 79566_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100005530.mm.1 | NM_001161338.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005530.mm.1 | NM_024480.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005530.mm.1 | TC1100000688.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416223_PM_at | - | - |
| Mouse Expression Set 430 | 1416223_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416223_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416223_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416223_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416223_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 925458 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6770601 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6770601 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006770601 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2734789 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2734789 | - | - |
| mouse-6 v1.1 (Illumina) | scl41519.8_316-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6770601 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41519.8_316-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6770601 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2734789 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2734789 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl41519.8_316-S | - | - |

Sh3bp5l
Genomic Map