Assay Details
| Gene Symbol: Sh3bp5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK122543.1 | - | 3 - 4 | 4218 | 61 | |
| AK144928.1 | - | 5 - 6 | 1013 | 61 | ||
| AK150129.1 | - | 5 - 6 | 1001 | 61 | ||
| AK156171.1 | - | 5 - 6 | 1054 | 61 | ||
| AK159202.1 | - | 5 - 6 | 1044 | 61 | ||
| AK167203.1 | - | 5 - 6 | 1128 | 61 | ||
| AK172333.1 | - | 5 - 6 | 1054 | 61 | ||
| BC003251.1 | - | 4 - 5 | 675 | 3500047 | 61 | |
| BC013661.1 | - | 1 - 2 | 146 | 61 | ||
| BC094418.1 | - | 5 - 6 | 1036 | 30702568 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3bp5l
Genomic Map