Assay Details
| Gene Symbol: Car13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024495.5 | NP_078771.1 | 1 - 2 | 122 | 104 | |
| GenBank mRNA | AF231123.1 | - | 1 - 2 | 72 | 104 | |
| AK010166.1 | - | 1 - 2 | 76 | 104 | ||
| AK150871.1 | - | 1 - 2 | 74 | 104 | ||
| AK150897.1 | - | 1 - 2 | 74 | 104 | ||
| AK151978.1 | - | 1 - 2 | 74 | 104 | ||
| AK152124.1 | - | 1 - 2 | 72 | 104 | ||
| AK153080.1 | - | 1 - 2 | 101 | 104 | ||
| AK153353.1 | - | 1 - 2 | 101 | 104 | ||
| AK162621.1 | - | 1 - 2 | 122 | 104 | ||
| BC064050.1 | - | 1 - 2 | 73 | 949505 | 104 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF231123.1,AK010166.1,AK150871.1,AK150897.1,AK151978.1,AK153080.1,AK153353.1,AK162621.1,BC064050.1,NM_024495.5 transcripts.
Set Membership: |
|

Car13
Genomic Map