Assay Details
| Gene Symbol: Car13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024495.5 | NP_078771.1 | 3 - 4 | 440 | 67 | |
| GenBank mRNA | AF231123.1 | - | 3 - 4 | 390 | 67 | |
| AK010166.1 | - | 3 - 4 | 394 | 67 | ||
| AK150871.1 | - | 3 - 4 | 392 | 67 | ||
| AK150897.1 | - | 3 - 4 | 392 | 67 | ||
| AK151519.1 | - | 3 - 4 | 378 | 67 | ||
| AK151978.1 | - | 3 - 4 | 392 | 67 | ||
| AK152124.1 | - | 3 - 4 | 389 | 67 | ||
| AK153080.1 | - | 3 - 4 | 419 | 67 | ||
| AK153258.1 | - | 3 - 4 | 380 | 67 | ||
| AK153353.1 | - | 3 - 4 | 419 | 67 | ||
| AK162621.1 | - | 3 - 4 | 440 | 67 | ||
| BC064050.1 | - | 3 - 4 | 391 | 949505 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car13
Genomic Map