Assay Details
| Gene Symbol: Car13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024495.5 | NP_078771.1 | 4 - 5 | 526 | 136 | |
| GenBank mRNA | AF231123.1 | - | 4 - 5 | 476 | 136 | |
| AK010166.1 | - | 4 - 5 | 480 | 136 | ||
| AK150871.1 | - | 4 - 5 | 478 | 136 | ||
| AK150897.1 | - | 4 - 5 | 478 | 136 | ||
| AK151519.1 | - | 4 - 5 | 464 | 136 | ||
| AK151978.1 | - | 4 - 5 | 478 | 136 | ||
| AK152124.1 | - | 4 - 5 | 475 | 136 | ||
| AK153080.1 | - | 4 - 5 | 505 | 136 | ||
| AK153258.1 | - | 4 - 5 | 466 | 136 | ||
| AK153353.1 | - | 4 - 5 | 505 | 136 | ||
| AK162621.1 | - | 4 - 5 | 526 | 136 | ||
| BC064050.1 | - | 4 - 5 | 477 | 949505 | 136 | |
More Information
Set Membership: |
|

Car13
Genomic Map