Assay Details
| Gene Symbol: Tmem222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025667.3 | NP_079943.2 | 2 - 3 | 341 | 86 | |
| GenBank mRNA | AK017503.1 | - | 2 - 3 | 337 | 86 | |
| AK079189.1 | - | 2 - 3 | 329 | 86 | ||
| AK160188.1 | - | 2 - 3 | 342 | 86 | ||
| BC037609.1 | - | 2 - 3 | 302 | 5356647 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem222
Genomic Map