Assay Details
| Gene Symbol: Tmem222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025667.3 | NP_079943.2 | 4 - 5 | 475 | 62 | |
| NR_110978.1 | 3 - 4 | 390 | 62 | |||
| GenBank mRNA | AK017503.1 | - | 4 - 5 | 471 | 62 | |
| AK079189.1 | - | 4 - 5 | 463 | 62 | ||
| AK160188.1 | - | 4 - 5 | 476 | 62 | ||
| BC037609.1 | - | 4 - 5 | 436 | 5356647 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem222
Genomic Map